3-Bromo-2-chloro-4-(p-tolyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

61408-500MG 61408-1G 61408-5G
77336-974EA 401.7 USD
77336-974 77355-480 77379-538
3-Bromo-2-chloro-4-(p-tolyl)pyridine ≥95%
3-Bromo-2-chloro-4-(p-tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₉BrClN
MW: 282.5635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760327
CAS Number: 2764729-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR