3-Bromo-2-chloro-4-(4-fluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

61407-500MG 61407-1G 61407-5G
77340-248EA 854.9 USD
77340-248 77355-478 77379-536
3-Bromo-2-chloro-4-(4-fluorophenyl)pyridine ≥97%
3-Bromo-2-chloro-4-(4-fluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₆BrClFN
MW: 286.5274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760326
CAS Number: 2764729-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR