3-(3-Bromo-2-chloropyridin-4-yl)benzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
61406-500MG
61406-1G
61406-5G
77340-228EA
845.63
USD
77340-228
77355-476
77379-534
3-(3-Bromo-2-chloropyridin-4-yl)benzaldehyde ≥97%
3-(3-Bromo-2-chloropyridin-4-yl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₂H₇BrClNO MW: 296.5470 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34760325 CAS Number: 2764729-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...