(4-Fluoro-[1,1'-biphenyl]-3,5-diyl)bis(methylsulphane) ≥97%

Supplier: AOBChem USA

Warning

61374-500MG 61374-1G 61374-5G
77338-782EA 517.06 USD
77338-782 77355-472 77379-528
(4-Fluoro-[1,1'-biphenyl]-3,5-diyl)bis(methylsulphane) ≥97%
(4-Fluoro-[1,1'-biphenyl]-3,5-diyl)bis(methylsulphane)

Precaución: for Research Use Only

Formula: C₁₄H₁₃FS₂
Storage Temperature: Ambient
MDL Number: MFCD34760298
CAS Number: 2764731-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR