(4-Fluoro-2'-methyl-[1,1'-biphenyl]-3,5-diyl)bis(methylsulphane) ≥97%

Supplier: AOBChem USA

Warning

61373-500MG 61373-1G 61373-5G
77338-022EA 459.38 USD
77338-022 77355-470 77379-526
(4-Fluoro-2'-methyl-[1,1'-biphenyl]-3,5-diyl)bis(methylsulphane) ≥97%
(4-Fluoro-2'-methyl-[1,1'-biphenyl]-3,5-diyl)bis(methylsulphane)

Precaución: for Research Use Only

Formula: C₁₅H₁₅FS₂
Storage Temperature: Ambient
MDL Number: MFCD34760297
CAS Number: 2764731-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR