2-Bromo-6-chloro-3-(difluoromethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

61371-500MG 61371-1G 61371-5G 61371-25G
77331-222EA 184.37 USD
77331-222 77347-248 77367-386 77388-906
2-Bromo-6-chloro-3-(difluoromethoxy)pyridine ≥97%
2-Bromo-6-chloro-3-(difluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrClF₂NO
MW: 258.4479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28733842
CAS Number: 1805576-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR