2-Bromo-6-chloro-3-isopropoxypyridine ≥95%

Supplier: AOBChem USA

Warning

61370-500MG 61370-1G 61370-5G 61370-25G
77331-220EA 182.31 USD
77331-220 77346-976 77367-384 77388-904
2-Bromo-6-chloro-3-isopropoxypyridine ≥95%
2-Bromo-6-chloro-3-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₉BrClNO
Storage Temperature: Ambient
MDL Number: MFCD34760295
CAS Number: 2807440-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR