2-Bromo-6-chloro-3-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

61368-500MG 61368-1G 61368-5G
77331-216EA 334.75 USD
77331-216 77355-468 77379-524
2-Bromo-6-chloro-3-ethoxypyridine ≥97%
2-Bromo-6-chloro-3-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇BrClNO
MW: 236.4935 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760293
CAS Number: 2755724-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR