1-Chloro-4-ethoxy-5-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61362-500MG 61362-1G 61362-5G
77331-212EA 314.15 USD
77331-212 77355-462 77379-520
1-Chloro-4-ethoxy-5-fluoro-2-methylbenzene ≥97%
1-Chloro-4-ethoxy-5-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClFO
MW: 188.6265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751513
CAS Number: 1805638-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR