(4-Fluoro-4'-methyl-[1,1'-biphenyl]-3,5-diyl)bis(methylsulphane) ≥97%

Supplier: AOBChem USA

Warning

61352-500MG 61352-1G 61352-5G
77331-210EA 321.36 USD
77331-210 77355-456 77379-514
(4-Fluoro-4'-methyl-[1,1'-biphenyl]-3,5-diyl)bis(methylsulphane) ≥97%
(4-Fluoro-4'-methyl-[1,1'-biphenyl]-3,5-diyl)bis(methylsulphane)

Precaución: for Research Use Only

Formula: C₁₅H₁₅FS₂
Storage Temperature: Ambient
CAS Number: 2918963-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR