4-Fluoro-3,5-bis(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61347-500MG 61347-1G 61347-5G
77336-646EA 390.37 USD
77336-646 77355-452 77379-510
4-Fluoro-3,5-bis(methylthio)benzaldehyde ≥97%
4-Fluoro-3,5-bis(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FOS₂
MW: 216.2955 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760283
CAS Number: 2807455-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR