Methyl 4-fluoro-3,5-bis(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

61345-500MG 61345-1G 61345-5G
77335-942EA 369.77 USD
77335-942 77355-448 77379-506
Methyl 4-fluoro-3,5-bis(methylthio)benzoate ≥97%
Methyl 4-fluoro-3,5-bis(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂S₂
MW: 246.3215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760281
CAS Number: 2901097-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR