4-Fluoro-3,5-bis(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

61344-500MG 61344-1G 61344-5G
77336-514EA 388.31 USD
77336-514 77355-446 77379-504
4-Fluoro-3,5-bis(methylthio)benzoic acid ≥97%
4-Fluoro-3,5-bis(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S₂
Storage Temperature: Ambient
MDL Number: MFCD34760280
CAS Number: 847862-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR