(5-Bromo-2-fluoro-1,3-phenylene)bis(methylsulphane) ≥97%

Supplier: AOBChem USA

Warning

61341-500MG 61341-1G 61341-5G 61341-25G
77331-206EA 180.25 USD
77331-206 77346-758 77367-378 77388-898
(5-Bromo-2-fluoro-1,3-phenylene)bis(methylsulphane) ≥97%
(5-Bromo-2-fluoro-1,3-phenylene)bis(methylsulphane)

Precaución: for Research Use Only

Formula: C₈H₈BrFS₂
MW: 267.1815 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760277
CAS Number: 2755722-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR