3-Ethoxy-2-fluoro-5-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61321-500MG 61321-1G 61321-5G 61321-25G
77331-196EA 168.92 USD
77331-196 77346-302 77367-370 77388-888
3-Ethoxy-2-fluoro-5-iodobenzoic acid ≥97%
3-Ethoxy-2-fluoro-5-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈FIO₃
MW: 310.0608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760259
CAS Number: 2090131-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR