3-Ethoxy-2-fluoro-5-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61319-500MG 61319-1G 61319-5G 61319-25G
77331-194EA 184.37 USD
77331-194 77347-170 77367-368 77388-886
3-Ethoxy-2-fluoro-5-iodobenzaldehyde ≥97%
3-Ethoxy-2-fluoro-5-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈FIO₂
Storage Temperature: Ambient
MDL Number: MFCD34760257
CAS Number: 2091531-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR