2-Chloro-4-fluoro-6-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

61308-500MG 61308-1G 61308-5G
77331-190EA 328.57 USD
77331-190 77355-432 77379-496
2-Chloro-4-fluoro-6-(methylthio)phenol ≥97%
2-Chloro-4-fluoro-6-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆ClFOS
MW: 192.6383 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760251
CAS Number: 2711476-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR