2,4-Dichloro-4'-fluoro-5-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61303-500MG 61303-1G 61303-5G
77339-038EA 545.9 USD
77339-038 77355-430 77379-494
2,4-Dichloro-4'-fluoro-5-methyl-1,1'-biphenyl ≥97%
2,4-Dichloro-4'-fluoro-5-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉Cl₂F
MW: 255.1149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760249
CAS Number: 2764729-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR