2,4-Dichloro-5-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

61298-500MG 61298-1G 61298-5G 61298-25G
77331-188EA 281.19 USD
77331-188 77355-428 77367-364 77388-882
2,4-Dichloro-5-methyl-1,1'-biphenyl ≥97%
2,4-Dichloro-5-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀Cl₂
Storage Temperature: Ambient
MDL Number: MFCD34760244
CAS Number: 2764731-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR