(E)-3-bromo-5-chloro-2-methoxybenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

61295-500MG 61295-1G 61295-5G 61295-25G
77331-186EA 267.8 USD
77331-186 77355-426 77367-362 77388-880
(E)-3-bromo-5-chloro-2-methoxybenzaldehyde oxime ≥97%
(E)-3-bromo-5-chloro-2-methoxybenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₈H₇BrClNO₂
MW: 264.5036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09707267
CAS Number: 938134-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR