1-Fluoro-5-iodo-2,4-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

61276-500MG 61276-1G 61276-25G 61276-5G
77331-180EA 299.73 USD
77331-180 77355-418 77388-874 77379-488
1-Fluoro-5-iodo-2,4-dimethoxybenzene ≥95%
1-Fluoro-5-iodo-2,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈FIO₂
MW: 282.0507 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547862
CAS Number: 879288-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR