2,3-Difluoro-1-(methoxymethyl)-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

61257-1G 61257-5G 61257-25G
77341-260EA 79.31 USD
77341-260 77367-352 77379-482
2,3-Difluoro-1-(methoxymethyl)-4-methylbenzene ≥95%
2,3-Difluoro-1-(methoxymethyl)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
MW: 172.1719 g/mol
Storage Temperature: Ambient
CAS Number: 2918947-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR