Methyl 2-iodo-4-methylthiophene-3-carboxylate ≥95%

Supplier: AOBChem USA

Warning

61248-500MG 61248-1G 61248-5G 61248-25G
77331-168EA 220.42 USD
77331-168 77355-406 77367-348 77388-862
Methyl 2-iodo-4-methylthiophene-3-carboxylate ≥95%
Methyl 2-iodo-4-methylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇IO₂S
MW: 282.0987 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760217
CAS Number: 2755722-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR