4-Bromo-N-methoxy-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

61241-1G 61241-5G 61241-25G
77341-572EA 93.73 USD
77341-572 77367-346 77379-478
4-Bromo-N-methoxy-3-methylbenzamide ≥97%
4-Bromo-N-methoxy-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.0852 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26802228
CAS Number: 222028-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR