Ethyl 4-chloro-2,3-difluoro-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

61240-500MG 61240-1G 61240-5G
77335-940EA 369.77 USD
77335-940 77355-404 77379-476
Ethyl 4-chloro-2,3-difluoro-5-methoxybenzoate ≥97%
Ethyl 4-chloro-2,3-difluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉ClF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34760216
CAS Number: 2764731-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR