3-Chloro-4-methoxy-2-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

61231-500MG 61231-1G 61231-5G 61231-25G
77331-164EA 264.71 USD
77331-164 77355-398 77367-344 77388-860
3-Chloro-4-methoxy-2-(methylthio)benzoic acid ≥97%
3-Chloro-4-methoxy-2-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃S
MW: 232.6839 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760211
CAS Number: 157929-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR