3-Ethyl-4-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

61213-500MG 61213-1G 61213-5G
77335-588EA 358.44 USD
77335-588 77355-394 77379-470
3-Ethyl-4-hydroxybenzaldehyde ≥97%
3-Ethyl-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD16997591
CAS Number: 105211-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR