2-Chloro-1-isopropoxy-4-phenoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61200-500MG 61200-1G 61200-5G
77331-158EA 322.39 USD
77331-158 77355-388 77379-464
2-Chloro-1-isopropoxy-4-phenoxybenzene ≥97%
2-Chloro-1-isopropoxy-4-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅ClO₂
MW: 262.7314 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760198
CAS Number: 2807441-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR