1-(Benzyloxy)-2-chloro-4-phenoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61199-500MG 61199-1G 61199-5G 61199-25G
77331-156EA 200.85 USD
77331-156 77355-386 77367-340 77388-854
1-(Benzyloxy)-2-chloro-4-phenoxybenzene ≥97%
1-(Benzyloxy)-2-chloro-4-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₉H₁₅ClO₂
MW: 310.7742 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760197
CAS Number: 2901103-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR