2-Chloro-1-ethoxy-4-phenoxybenzene ≥95%

Supplier: AOBChem USA

Warning

61198-500MG 61198-1G 61198-5G 61198-25G
77331-154EA 217.33 USD
77331-154 77355-384 77367-338 77388-852
2-Chloro-1-ethoxy-4-phenoxybenzene ≥95%
2-Chloro-1-ethoxy-4-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClO₂
MW: 248.7048 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760196
CAS Number: 2901109-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR