2-Chloro-1-methoxy-4-phenoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61197-500MG 61197-1G 61197-25G 61197-5G
77331-152EA 289.43 USD
77331-152 77355-382 77388-850 77379-462
2-Chloro-1-methoxy-4-phenoxybenzene ≥97%
2-Chloro-1-methoxy-4-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34760195
CAS Number: 2918858-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR