1-Bromo-3-chloro-2-methoxy-5-phenoxybenzene ≥95%

Supplier: AOBChem USA

Warning

61196-500MG 61196-1G 61196-5G
77337-962EA 455.26 USD
77337-962 77355-380 77379-460
1-Bromo-3-chloro-2-methoxy-5-phenoxybenzene ≥95%
1-Bromo-3-chloro-2-methoxy-5-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrClO₂
MW: 313.5743 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760194
CAS Number: 2901065-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR