2-Bromo-6-chloro-4-phenoxyphenol ≥95%

Supplier: AOBChem USA

Warning

61195-500MG 61195-1G 61195-5G
77331-150EA 347.11 USD
77331-150 77355-378 77379-458
2-Bromo-6-chloro-4-phenoxyphenol ≥95%
2-Bromo-6-chloro-4-phenoxyphenol

Precaución: for Research Use Only

Formula: C₁₂H₈BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD34760193
CAS Number: 2807467-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR