2-Bromo-1-methoxy-4-phenoxybenzene ≥97%

Supplier: AOBChem USA

Warning

61192-1G 61192-5G 61192-25G
77340-774EA 60.77 USD
77340-774 77367-336 77379-456
2-Bromo-1-methoxy-4-phenoxybenzene ≥97%
2-Bromo-1-methoxy-4-phenoxybenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD34760191
CAS Number: 58765-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR