5-Cyclopentyl-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

61155-500MG 61155-1G 61155-5G
77340-038EA 722.03 USD
77340-038 77355-368 77379-442
5-Cyclopentyl-2-methoxybenzoic acid ≥97%
5-Cyclopentyl-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD34760177
CAS Number: 1784029-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR