(5-Cyclopentyl-2-methoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

61153-500MG 61153-1G 61153-5G
77340-382EA 979.53 USD
77340-382 77355-366 77379-440
(5-Cyclopentyl-2-methoxyphenyl)boronic acid ≥95%
(5-Cyclopentyl-2-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD28167263
CAS Number: 1134263-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR