2-Bromo-4-cyclopentyl-1-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

61147-500MG 61147-1G 61147-5G
77338-048EA 460.41 USD
77338-048 77355-362 77379-436
2-Bromo-4-cyclopentyl-1-methoxybenzene ≥95%
2-Bromo-4-cyclopentyl-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO
MW: 255.1509 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760173
CAS Number: 59216-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR