5-(2,4-Dimethoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61130-500MG 61130-1G 61130-25G 61130-5G
77331-140EA 294.58 USD
77331-140 77355-360 77388-844 77379-432
5-(2,4-Dimethoxyphenyl)thiazol-2-amine ≥97%
5-(2,4-Dimethoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂S
MW: 236.2901 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01681565
CAS Number: 22035-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR