5-(4-Fluoro-2-isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61126-500MG 61126-1G 61126-25G 61126-5G
77331-136EA 305.91 USD
77331-136 77355-352 77388-842 77379-424
5-(4-Fluoro-2-isopropoxyphenyl)thiazol-2-amine ≥97%
5-(4-Fluoro-2-isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃FN₂OS
MW: 252.3078 g/mol
Storage Temperature: Ambient
CAS Number: 2807445-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR