5-(3-Fluoro-2-(methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

61125-500MG 61125-1G 61125-5G
77337-176EA 408.91 USD
77337-176 77355-350 77379-422
5-(3-Fluoro-2-(methylthio)phenyl)thiazol-2-amine ≥97%
5-(3-Fluoro-2-(methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S₂
MW: 240.3202 g/mol
Storage Temperature: Ambient
CAS Number: 2807450-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR