2-Chloro-1,3,5-trifluoro-4-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

61114-500MG 61114-1G 61114-5G 61114-25G
77331-134EA 128.75 USD
77331-134 77344-272 77367-330 77379-420
2-Chloro-1,3,5-trifluoro-4-iodobenzene ≥97%
2-Chloro-1,3,5-trifluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆HClF₃I
Storage Temperature: Ambient
MDL Number: MFCD34760145
CAS Number: 2757731-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR