1-Chloro-2,3,4-trifluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

61093-1G 61093-5G 61093-25G
77341-630EA 96.82 USD
77341-630 77367-322 77379-416
1-Chloro-2,3,4-trifluoro-5-iodobenzene ≥97%
1-Chloro-2,3,4-trifluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆HClF₃I
MW: 292.4248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760134
CAS Number: 2374235-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR