(5-Chloro-2,3,4-trifluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

61092-500MG 61092-1G 61092-5G 61092-25G
77331-128EA 100.94 USD
77331-128 77342-956 77367-320 77379-414
(5-Chloro-2,3,4-trifluorophenyl)(methyl)sulfane ≥97%
(5-Chloro-2,3,4-trifluorophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₇H₄ClF₃S
MW: 212.6198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760133
CAS Number: 2764732-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR