1-(5-Chloro-2,3,4-trifluorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

61090-500MG 61090-1G 61090-5G 61090-25G
77331-126EA 124.63 USD
77331-126 77344-064 77367-318 77379-412
1-(5-Chloro-2,3,4-trifluorophenyl)ethanone ≥97%
1-(5-Chloro-2,3,4-trifluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
MW: 208.5649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760132
CAS Number: 2764729-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR