Ethyl 5-chloro-2,3,4-trifluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

61087-500MG 61087-1G 61087-5G 61087-25G
77331-124EA 247.2 USD
77331-124 77355-346 77367-316 77388-836
Ethyl 5-chloro-2,3,4-trifluorobenzoate ≥97%
Ethyl 5-chloro-2,3,4-trifluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD14698201
CAS Number: 1214332-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR