Methyl 5-chloro-2,3,4-trifluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

61086-500MG 61086-1G 61086-5G 61086-25G
77331-122EA 147.29 USD
77331-122 77345-232 77367-314 77388-834
Methyl 5-chloro-2,3,4-trifluorobenzoate ≥95%
Methyl 5-chloro-2,3,4-trifluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD14698011
CAS Number: 1214335-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR