(6-Bromo-2,3,4-trifluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

61081-500MG 61081-1G 61081-5G 61081-25G
77331-116EA 126.69 USD
77331-116 77344-174 77367-308 77379-408
(6-Bromo-2,3,4-trifluorophenyl)(methyl)sulfane ≥97%
(6-Bromo-2,3,4-trifluorophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₇H₄BrF₃S
MW: 257.0708 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760129
CAS Number: 2764728-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR