Methyl 6-methoxy-4-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

61072-500MG 61072-1G 61072-5G
77340-010EA 712.76 USD
77340-010 77355-342 77379-404
Methyl 6-methoxy-4-methylnicotinate ≥97%
Methyl 6-methoxy-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD18910323
CAS Number: 1355229-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR