Methyl(2,3,4-trifluoro-5-iodophenyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

61071-500MG 61071-1G 61071-5G 61071-25G
77331-110EA 122.57 USD
77331-110 77343-954 77367-304 77379-402
Methyl(2,3,4-trifluoro-5-iodophenyl)sulfane ≥97%
Methyl(2,3,4-trifluoro-5-iodophenyl)sulphane

Precaución: for Research Use Only

Formula: C₇H₄F₃IS
MW: 304.0713 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760127
CAS Number: 2764728-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR