Methyl 4-fluoro-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

61057-500MG 61057-1G 61057-25G 61057-5G
77331-108EA 305.91 USD
77331-108 77355-338 77388-828 77379-398
Methyl 4-fluoro-3-(methylthio)benzoate ≥97%
Methyl 4-fluoro-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
MW: 200.2299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489118
CAS Number: 1823098-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR